Assay Details
| Gene Symbol: CDC26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302202.1 | NP_001289131.1 | 2 | 224 | 116 | |
| GenBank mRNA | BX934329.2 | - | 2 | 162 | 116 | |
| CN237067.1 | - | 2 | 224 | 116 | ||
More Information
Set Membership: |
|

CDC26
Genomic Map