Assay Details
| Gene Symbol: NMUR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318436.1 | NP_001305365.1 | 3 | 1720 | 100 | |
| XM_015291493.2 | XP_015146979.1 | 4 | 100 | |||
| XM_015291494.2 | XP_015146980.1 | 4 | 100 | |||
| GenBank mRNA | HQ833216.1 | - | 3 | 1720 | 100 | |
More Information
Set Membership: |
|

NMUR1
Genomic Map