Assay Details
| Gene Symbol: GDF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205211.1 | NP_990542.1 | 2 | 1220 | 59 | |
| GenBank mRNA | U55871.1 | - | 2 | 1244 | 59 | |
| U73003.1 | - | 2 | 1112 | 59 | ||
More Information
Set Membership: |
|

GDF3
Genomic Map