Assay Details
| Gene Symbol: SLIRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001235248.5 | XP_001235249.1 | 3 - 4 | 273 | 80 | |
| GenBank mRNA | CR352594.1 | - | 5 - 6 | 645 | 80 | |
More Information
Set Membership: |
|

SLIRP
Genomic Map