Assay Details
| Gene Symbol: SETD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233555.5 | XP_001233556.3 | 4 - 5 | 764 | 75 | |
| XM_004937248.3 | XP_004937305.2 | 4 - 5 | 816 | 75 | ||
| XM_025144857.1 | XP_025000625.1 | 4 - 5 | 725 | 75 | ||
| GenBank mRNA | CR352813.1 | - | 4 - 5 | 450 | 75 | |
| CR523214.1 | - | 4 - 5 | 751 | 75 | ||
More Information
Set Membership: |
|

SETD9
Genomic Map