Assay Details
| Gene Symbol: ZC3H12D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004935603.3 | XP_004935660.1 | 4 - 5 | 1628 | 101 | |
| XM_015284274.1 | XP_015139760.1 | 3 - 4 | 1007 | 101 | ||
| XM_025149064.1 | XP_025004832.1 | 3 - 4 | 2981 | 101 | ||
| XM_025149065.1 | XP_025004833.1 | 3 - 4 | 1524 | 101 | ||
| XM_025149066.1 | XP_025004834.1 | 3 - 4 | 977 | 101 | ||
| GenBank mRNA | CR353299.1 | - | 2 - 3 | 243 | 101 | |
More Information
Set Membership: |
|

ZC3H12D
Genomic Map