Assay Details
| Gene Symbol: PQLC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025149413.1 | XP_025005181.1 | 7 | 2096 | 113 | |
| XM_419953.4 | XP_419953.2 | 7 | 2016 | 113 | ||
| GenBank mRNA | CR353570.1 | - | 1 | 722 | 113 | |
More Information
Set Membership: |
|

PQLC3
Genomic Map