Assay Details
| Gene Symbol: TBC1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004949608.3 | XP_004949665.1 | 5 - 6 | 1126 | 63 | |
| XM_004949609.3 | XP_004949666.1 | 5 - 6 | 1092 | 63 | ||
| XM_015280669.2 | XP_015136155.1 | 6 - 7 | 1717 | 63 | ||
| XM_015280670.2 | XP_015136156.1 | 5 - 6 | 960 | 63 | ||
| XM_015280671.2 | XP_015136157.1 | 5 - 6 | 1582 | 63 | ||
| XM_015280672.2 | XP_015136158.1 | 5 - 6 | 665 | 63 | ||
| GenBank mRNA | CR353814.1 | - | 3 - 4 | 410 | 63 | |
More Information
Set Membership: |
|

TBC1D2
Genomic Map