Assay Details
| Gene Symbol: XPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015293355.2 | XP_015148841.1 | 3 - 4 | 465 | 107 | |
| XM_414379.6 | XP_414379.3 | 3 - 4 | 465 | 107 | ||
| GenBank mRNA | CR354014.1 | - | 523 | 107 | ||
More Information
Set Membership: |
|

XPC
Genomic Map