Assay Details
| Gene Symbol: IFI30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_418246.5 | XP_418246.4 | 1 - 2 | 454 | 80 | |
| GenBank mRNA | CR354229.1 | - | 1 - 2 | 141 | 80 | |
| JX843734.1 | - | 1 - 2 | 124 | 80 | ||
More Information
Set Membership: |
|

IFI30
Genomic Map