Assay Details
| Gene Symbol: RNF168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233497.5 | XP_001233498.1 | 7 | 1543 | 93 | |
| XM_004936998.3 | XP_004937055.1 | 7 | 1873 | 93 | ||
| XM_015277130.2 | XP_015132616.1 | 7 | 1682 | 93 | ||
| GenBank mRNA | CR385404.1 | - | 1 | 126 | 93 | |
More Information
Set Membership: |
|

RNF168
Genomic Map