Assay Details
| Gene Symbol: VSTM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421654.5 | XP_421654.3 | 8 | 2429 | 92 | |
| GenBank mRNA | CR385514.1 | - | 1 | 621 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VSTM4| Gene Symbol: VSTM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421654.5 | XP_421654.3 | 8 | 2429 | 92 | |
| GenBank mRNA | CR385514.1 | - | 1 | 621 | 92 | |
Set Membership: |
|