Assay Details
| Gene Symbol: GABPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642687.4 | XP_003642735.1 | 5 - 6 | 600 | 79 | |
| XM_015280044.2 | XP_015135530.1 | 5 - 6 | 628 | 79 | ||
| XM_025143613.1 | XP_024999381.1 | 5 - 6 | 628 | 79 | ||
| XM_025143614.1 | XP_024999382.1 | 5 - 6 | 629 | 79 | ||
| GenBank mRNA | CR386142.1 | - | 1 - 2 | 205 | 79 | |
More Information
Set Membership: |
|

GABPB2
Genomic Map