Assay Details
| Gene Symbol: C4HCXorf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278635.2 | XP_015134121.1 | 1 - 2 | 5950 | 95 | |
| XM_015278636.2 | XP_015134122.1 | 1 - 2 | 5950 | 95 | ||
| XM_025150265.1 | XP_025006033.1 | 1 - 2 | 5950 | 95 | ||
| GenBank mRNA | CR386396.1 | - | 1 - 2 | 143 | 95 | |
More Information
Set Membership: |
|

C4HCXorf57
Genomic Map