Assay Details
| Gene Symbol: KHDRBS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025149087.1 | XP_025004855.1 | 9 | 1625 | 95 | |
| XM_426201.6 | XP_426201.2 | 9 | 1640 | 95 | ||
| GenBank mRNA | CR386489.1 | - | 1 | 120 | 95 | |
| CR406447.1 | - | 4 | 433 | 95 | ||
More Information
Set Membership: |
|

KHDRBS2
Genomic Map