Assay Details
| Gene Symbol: WISP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004947031.3 | XP_004947088.3 | 2 - 3 | 99 | ||
| XM_417370.6 | XP_417370.6 | 2 - 3 | 99 | |||
| GenBank mRNA | CR386673.1 | - | 1 - 2 | 123 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004947031.3 transcripts.
Set Membership: |
|

WISP2
Genomic Map