Assay Details
| Gene Symbol: SEPT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015277915.2 | XP_015133401.1 | 2 - 3 | 233 | 131 | |
| XM_025146690.1 | XP_025002458.1 | 3 - 4 | 223 | 131 | ||
| GenBank mRNA | CR386973.1 | - | 2 - 3 | 158 | 131 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025146690.1 transcripts.
Set Membership: |
|

SEPT14
Genomic Map