Assay Details
| Gene Symbol: YIPF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420160.6 | XP_420160.3 | 5 - 6 | 605 | 93 | |
| GenBank mRNA | CR387744.1 | - | 1 - 2 | 49 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

YIPF6| Gene Symbol: YIPF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420160.6 | XP_420160.3 | 5 - 6 | 605 | 93 | |
| GenBank mRNA | CR387744.1 | - | 1 - 2 | 49 | 93 | |
Set Membership: |
|