Assay Details
| Gene Symbol: ZNF827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004940959.3 | XP_004941016.1 | 14 | 10675 | 105 | |
| XM_015276711.2 | XP_015132197.1 | 14 | 4605 | 105 | ||
| XM_025149914.1 | XP_025005682.1 | 14 | 10672 | 105 | ||
| XM_025149915.1 | XP_025005683.1 | 12 | 10019 | 105 | ||
| GenBank mRNA | CR389399.1 | - | 1 | 512 | 105 | |
More Information
Set Membership: |
|

ZNF827
Genomic Map