Assay Details
| Gene Symbol: CHPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_423717.6 | XP_423717.3 | 3 - 4 | 1367 | 61 | |
| GenBank mRNA | CR389631.1 | - | 2 - 3 | 564 | 61 | |
More Information
Set Membership: |
|

CHPF2
Genomic Map