Assay Details
| Gene Symbol: DBF4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234425.5 | XP_001234426.3 | 11 | 1738 | 77 | |
| XM_004948479.3 | XP_004948536.1 | 11 | 1741 | 77 | ||
| XM_015299264.2 | XP_015154750.1 | 12 | 1614 | 77 | ||
| GenBank mRNA | CR389846.1 | - | 1 | 519 | 77 | |
More Information
Set Membership: |
|

DBF4B
Genomic Map