Assay Details
| Gene Symbol: PRDM12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415465.5 | XP_415465.2 | 5 | 1382 | 96 | |
| GenBank mRNA | CR390550.1 | - | 1 | 325 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRDM12| Gene Symbol: PRDM12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415465.5 | XP_415465.2 | 5 | 1382 | 96 | |
| GenBank mRNA | CR390550.1 | - | 1 | 325 | 96 | |
Set Membership: |
|