Assay Details
| Gene Symbol: DNAJC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004950038.3 | XP_004950095.3 | 2 - 3 | 82 | ||
| XM_025145645.1 | XP_025001413.1 | 2 - 3 | 82 | |||
| XM_428430.6 | XP_428430.5 | 2 - 3 | 82 | |||
| GenBank mRNA | CR390686.1 | - | 1 - 2 | 685 | 82 | |
| CR524062.1 | - | 2 - 3 | 673 | 82 | ||
More Information
Set Membership: |
|

DNAJC22
Genomic Map