Assay Details
| Gene Symbol: OCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004938466.3 | XP_004938523.1 | 21 - 22 | 2928 | 88 | |
| XM_025146634.1 | XP_025002402.1 | 20 - 21 | 2687 | 88 | ||
| XM_025146635.1 | XP_025002403.1 | 20 - 21 | 2279 | 88 | ||
| XM_025146636.1 | XP_025002404.1 | 19 - 20 | 2354 | 88 | ||
| GenBank mRNA | AB617822.1 | - | 17 - 18 | 1806 | 88 | |
| AB617823.1 | - | 17 - 18 | 1806 | 88 | ||
More Information
Set Membership: |
|

OCA2
Genomic Map