Assay Details
| Gene Symbol: RPS26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015300309.2 | XP_015155795.1 | 2 - 3 | 202 | 78 | |
| GenBank mRNA | AJ719518.1 | - | 2 - 3 | 201 | 78 | |
More Information
Set Membership: |
|

RPS26
Genomic Map