Assay Details
| Gene Symbol: WNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015273182.2 | XP_015128668.1 | 3 - 4 | 1455 | 67 | |
| GenBank mRNA | AY655699.1 | - | 2 - 3 | 526 | 67 | |
| AY753286.1 | - | 2 - 3 | 497 | 67 | ||
More Information
Set Membership: |
|

WNT1
Genomic Map