Assay Details
| Gene Symbol: BOLA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297467.2 | XP_015152953.1 | 2 - 3 | 173 | 94 | |
| GenBank mRNA | CR391436.1 | - | 1 - 2 | 108 | 94 | |
More Information
Set Membership: |
|

BOLA3
Genomic Map