Assay Details
| Gene Symbol: BEND4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015285472.2 | XP_015140958.1 | 5 | 4398 | 90 | |
| XM_420730.6 | XP_420730.4 | 5 | 4467 | 90 | ||
| GenBank mRNA | CR391527.1 | - | 1 | 418 | 90 | |
More Information
Set Membership: |
|

BEND4
Genomic Map