Assay Details
| Gene Symbol: BSND | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015291071.2 | XP_015146557.1 | 3 - 4 | 581 | 115 | |
| GenBank mRNA | CR391599.1 | - | 1 - 2 | 91 | 115 | |
More Information
Set Membership: |
|

BSND
Genomic Map