Assay Details
| Gene Symbol: PKDCCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421361.6 | XP_421361.3 | 4 - 5 | 1228 | 106 | |
| GenBank mRNA | CR405754.1 | - | 2 - 3 | 207 | 106 | |
More Information
Set Membership: |
|

PKDCCB
Genomic Map