Assay Details
| Gene Symbol: PKDCCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421361.6 | XP_421361.3 | 5 - 6 | 1340 | 149 | |
| GenBank mRNA | CR405754.1 | - | 3 - 4 | 319 | 149 | |
More Information
Set Membership: |
|

PKDCCB
Genomic Map