Assay Details
| Gene Symbol: GABRD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234040.4 | XP_001234041.2 | 3 - 4 | 739 | 75 | |
| XM_004947393.3 | XP_004947450.1 | 3 - 4 | 737 | 75 | ||
| XM_015297107.2 | XP_015152593.1 | 3 - 4 | 240 | 75 | ||
| GenBank mRNA | CR406266.1 | - | 3 - 4 | 247 | 75 | |
More Information
Set Membership: |
|

GABRD
Genomic Map