Assay Details
| Gene Symbol: ETNK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015299131.2 | XP_015154617.1 | 1280 | 105 | ||
| XM_025143869.1 | XP_024999637.1 | 1155 | 105 | |||
| XM_424313.6 | XP_424313.5 | 1375 | 105 | |||
| GenBank mRNA | CR406747.1 | - | 115 | 105 | ||
More Information
Set Membership: |
|

ETNK2
Genomic Map