Assay Details
| Gene Symbol: PURG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015276629.2 | XP_015132115.1 | 2 | 2601 | 101 | |
| GenBank mRNA | CR406959.1 | - | 1 | 214 | 101 | |
| CR407259.1 | - | 1 | 169 | 101 | ||
More Information
Set Membership: |
|

PURG
Genomic Map