Assay Details
| Gene Symbol: ZNF704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004939931.3 | XP_004939988.1 | 9 | 3138 | 120 | |
| XM_015282859.2 | XP_015138345.2 | 9 | 2693 | 120 | ||
| XM_025148016.1 | XP_025003784.1 | 8 | 2593 | 120 | ||
| XM_418305.6 | XP_418305.3 | 9 | 2590 | 120 | ||
| GenBank mRNA | CR407016.1 | - | 1 | 517 | 120 | |
More Information
Set Membership: |
|

ZNF704
Genomic Map