Assay Details
| Gene Symbol: NPTXR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155299.1 | XP_025011067.1 | 5 | 8135 | 62 | |
| GenBank mRNA | CR407152.1 | - | 1 | 826 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NPTXR| Gene Symbol: NPTXR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155299.1 | XP_025011067.1 | 5 | 8135 | 62 | |
| GenBank mRNA | CR407152.1 | - | 1 | 826 | 62 | |
Set Membership: |
|