Assay Details
| Gene Symbol: LOC100857191 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642981.4 | XP_003643029.1 | 2 - 3 | 242 | 112 | |
| GenBank mRNA | CR522995.1 | - | 2 - 3 | 224 | 112 | |
| MG066604.1 | - | 2 - 3 | 421 | 112 | ||
More Information
Set Membership: |
|

LOC100857191
Genomic Map