Assay Details
| Gene Symbol: ACSS1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025149029.1 | XP_025004797.1 | 11 - 12 | 1561 | 79 | |
| XM_415011.6 | XP_415011.5 | 13 - 14 | 3240 | 79 | ||
| GenBank mRNA | CR523173.1 | - | 1 - 2 | 57 | 79 | |
More Information
Set Membership: |
|

ACSS1A
Genomic Map