Assay Details
| Gene Symbol: CECR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004937949.3 | XP_004938006.1 | 8 | 4451 | 62 | |
| GenBank mRNA | CR523452.1 | - | 1 | 924 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CECR5| Gene Symbol: CECR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004937949.3 | XP_004938006.1 | 8 | 4451 | 62 | |
| GenBank mRNA | CR523452.1 | - | 1 | 924 | 62 | |
Set Membership: |
|