Assay Details
| Gene Symbol: SCFD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233405.5 | XP_001233406.4 | 9 | 3025 | 84 | |
| XM_015285524.2 | XP_015141010.1 | 9 | 3023 | 84 | ||
| GenBank mRNA | CR523759.1 | - | 1 | 1016 | 84 | |
More Information
Set Membership: |
|

SCFD2
Genomic Map