Assay Details
| Gene Symbol: SLC30A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015283896.2 | XP_015139382.1 | 4 | 3170 | 115 | |
| XM_015283897.2 | XP_015139383.1 | 4 | 3164 | 115 | ||
| GenBank mRNA | CR523791.1 | - | 1 | 813 | 115 | |
More Information
Set Membership: |
|

SLC30A10
Genomic Map