Assay Details
| Gene Symbol: SERTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278404.2 | XP_015133890.2 | 1 - 2 | 1037 | 75 | |
| XM_025146827.1 | XP_025002595.1 | 1 - 2 | 637 | 75 | ||
| XM_417096.6 | XP_417096.4 | 1 - 2 | 48 | 75 | ||
| GenBank mRNA | CR524398.1 | - | 1 - 2 | 42 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_417096.6 transcripts.
Set Membership: |
|

SERTM1
Genomic Map