Assay Details
| Gene Symbol: LOC428277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_425837.6 | XP_425837.4 | 7 - 8 | 3471 | 101 | |
| GenBank mRNA | EU250015.1 | - | 7 - 8 | 721 | 101 | |
| FR746110.1 | - | 7 - 8 | 847 | 101 | ||
More Information
Set Membership: |
|

LOC428277
Genomic Map