Assay Details
| Gene Symbol: LOC428277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_425837.6 | XP_425837.4 | 9 - 10 | 3591 | 65 | |
| GenBank mRNA | EU250015.1 | - | 9 - 10 | 841 | 65 | |
| FR746110.1 | - | 9 - 10 | 967 | 65 | ||
More Information
Set Membership: |
|

LOC428277
Genomic Map