Assay Details
| Gene Symbol: E2F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297719.2 | XP_015153205.2 | 3 - 4 | 1136 | 79 | |
| GenBank mRNA | FJ770467.1 | - | 1 - 2 | 218 | 79 | |
More Information
Set Membership: |
|

E2F2
Genomic Map