Assay Details
| Gene Symbol: SLC25A30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015276591.2 | XP_015132077.1 | 3 - 4 | 532 | 86 | |
| XM_417040.6 | XP_417040.3 | 3 - 4 | 524 | 86 | ||
More Information
Set Membership: |
|

SLC25A30
Genomic Map