Assay Details
| Gene Symbol: EPHX1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419497.6 | XP_419497.1 | 8 - 9 | 1221 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPHX1L| Gene Symbol: EPHX1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419497.6 | XP_419497.1 | 8 - 9 | 1221 | 112 | |
Set Membership: |
|