Assay Details
| Gene Symbol: GSK3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004938179.3 | XP_004938236.1 | 11 | 3674 | 97 | |
| XM_416557.6 | XP_416557.4 | 12 | 3713 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GSK3A| Gene Symbol: GSK3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004938179.3 | XP_004938236.1 | 11 | 3674 | 97 | |
| XM_416557.6 | XP_416557.4 | 12 | 3713 | 97 | ||
Set Membership: |
|