Assay Details
| Gene Symbol: NLK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015295865.2 | XP_015151351.1 | 9 - 10 | 1830 | 73 | |
| XM_015295866.2 | XP_015151352.1 | 9 - 10 | 1830 | 73 | ||
More Information
Set Membership: |
|

NLK
Genomic Map