Assay Details
| Gene Symbol: STEAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004939275.3 | XP_004939332.1 | 5 | 5101 | 123 | |
| XM_015281967.2 | XP_015137453.1 | 5 | 4581 | 123 | ||
| XM_015281968.2 | XP_015137454.1 | 5 | 4770 | 123 | ||
| XM_025148419.1 | XP_025004187.1 | 5 | 6420 | 123 | ||
More Information
Set Membership: |
|

STEAP2
Genomic Map